Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial, Cap and Septa Unassembled Component Kits with FEP/Natural Rubber Septa
SDP
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Supplier:  DWK Life Sciences 091020NT
 View more versions of this product
Catalog No. 15-111-155
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Yellow
12 mm
No
FEP, Natural Rubber
MicroLiter
Screw Cap
9 mm
Clear
Borosilicate Glass (Type 1)
2 mL
100
Polypropylene
32 mm
No
Recommendations
Recommendations