Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with Open Tops
SDP

MicroLiter's 9mm autosampler screw-thread vial caps with open tops are available in multiple colors.

Supplier:  DWK Life Sciences 090000G

 View more versions of this product

Catalog No. 15-111-008


Add to Cart

Description

Description

For use with MicroLiter 9mm Screwthread Autosample Vial Septa sold separately.
Specifications

Specifications

Green
None
Screw Cap
9 mm Autosampler Vials - MicroLiter
Polypropylene
100
9 mm
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit