Learn More
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with PTFE/Silicone Septa
Available in multiple colors and package sizes.
Manufacturer:  DWK Life Sciences 090034B
 View more versions of this product
Catalog No. 15-111-084
Add to cart
Specifications
Specifications
Blue
Polypropylene
No
PTFE/Silicone
PTFE/Silicone
9mm Cap
Screw Thread
9mm
Screw Thread
9mm Autosampler Vials - MicroLiter
100