Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vials With Insert
SDP

MicroLiter 9mm autosampler screwthread vials with inserts are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.

Supplier:  DWK Life Sciences 091000101

 View more versions of this product

Catalog No. 15-111-130


Explore available promotions

Description

Description

Performance equal to an 11 mm crimp-top vial. Inserts available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Borosilicate Glass (Type 1)
Glass, Conical with Pulled Point
32 mm
9 mm
9 mm Screw Thread Caps - MicroLiter
12 mm
350 μL
Untreated
100
No
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.