Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MicroLIVE Bacteria Cultures
Unique culture system design for easy observation of bacteriasimply squeeze, shake, and dispense.
Specifications
Product Type | Microlive Bacteria Culture |
---|---|
Quantity | Each |
For Use With (Application) | Easy observation of bacteria |
Includes | Agar slant for optional subculturing along with an informative key card |
Packaging | Ampules |
Includes: Informative key card with instructions for use and an overview of the structure and function of each organism.
Catalog Number | Mfr. No. | Scientific Name | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Scientific Name | Price | Quantity & Availability | |||||
S65032
![]() |
Neo/Sci Corporation
800012 |
E.coli |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Ampules require no refrigeration
- Can be stored at room temperature for up to 18 months prior to use
- Dispensing tube reduces student contact with the organism
- Economical design eliminates need for additional growth media or pipets
- Cultures require no incubation and can be used immediately
Specifications
Microlive Bacteria Culture | |
Easy observation of bacteria | |
Ampules |
Each | |
Agar slant for optional subculturing along with an informative key card | |
Room Temperature |
Safety and Handling
ShelfLife : 18 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title