Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
GREENER_CHOICE

Catalog No. p-7075451
Click to view available options
Color:
Blue, Clear Bottom
Clear
Clear, Blue
Clear, Green
Clear, Red
Clear, Yellow
Green, Clear Bottom
Red, Clear Bottom
Binding Property:
High Binding
Medium Binding

For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Individual wells can be pipetted without leaving residue
  • Flat bottom enables precise optical measurements
  • Suitable for transmission measurements
  • Specifica
TRUSTED_SUSTAINABILITY

Specifications

Breakapart Single break
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Packaging Inner bags of 10
Binding Type Medium Binding
Color Red, Clear Bottom
Product Type 96-well Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon
Well Shape Curved Edges, Flat Bottom (C-well)
Dnase Free Yes
For Use With (Application) Colorimetric immunoassay
Quantity 100/Cs.
RNase Free Yes
Dimensions (L x W x H) 127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.