Thermo Scientific™ Microm™ HM 550 Cryostats Blade Carriers
Use Thermo Scientific™ Microm™ HM 550 Blade Carriers with Richard-Allan Scientific™ Cryostats.
Manufacturer: Thermo Scientific™ 705720
 View more versions of this product
Catalog No. 70-572-0
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Description Blade Holder
    For Use With HM 560
    Use Thermo Scientific™ Microm™ HM 550 Blade Carriers with Richard-Allan Scientific™ Cryostats.
    Standard Knife Carrier:
    • Allows for insertion of standard knives, with clearance angle adjustment
    • Can be moved horizontally on precision dovetail guide with quick clamp system
    • Includes anti-roll plate
    Disposable Blade Carrier EC:
    • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
    • Can be moved on precision dovetail guide with quick clamp plate
    • Includes anti-roll plate
    Disposable Blade Carrier EV:
    • Use with vacutome system
    • Allows for insertion of high- and low-profile disposable blades
    • Includes a vacuum anti-roll hood for stretching and disposing of section waste
    • Features a slot that is opened and closed by a vacuum-controlled flap
    Magnetic Disposable Blade Carrier MV:
    • Use with vacutome system
    • Allows for insertion of disposable blades
    • Includes a vacuum anti-roll hood for stretching and disposing of section waste
    • Features a slot that is opened and closed by a vacuum-controlled flap
  • Description & Specifications
Related Products
Product Content Feedback