Thermo Scientific™ Microm™ HM 550 Cryostats Blade Carriers Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Microm™ HM 550 Blade Carriers with Richard-Allan Scientific™ Cryostats.
Manufacturer: Thermo Scientific™ 705580
 View more versions of this product
Catalog No. 14-071-09
  Check Availability
Add to Cart
    Specifications
    Description Disposable Blade Carrier EV
    For Use With HM 550 and HM 560
    Use Thermo Scientific™ Microm™ HM 550 Blade Carriers with Richard-Allan Scientific™ Cryostats.
    Standard Knife Carrier:
    • Allows for insertion of standard knives, with clearance angle adjustment
    • Can be moved horizontally on precision dovetail guide with quick clamp system
    • Includes anti-roll plate
    Disposable Blade Carrier EC:
    • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
    • Can be moved on precision dovetail guide with quick clamp plate
    • Includes anti-roll plate
    Disposable Blade Carrier EV:
    • Use with vacutome system
    • Allows for insertion of high- and low-profile disposable blades
    • Includes a vacuum anti-roll hood for stretching and disposing of section waste
    • Features a slot that is opened and closed by a vacuum-controlled flap
    Magnetic Disposable Blade Carrier MV:
    • Use with vacutome system
    • Allows for insertion of disposable blades
    • Includes a vacuum anti-roll hood for stretching and disposing of section waste
    • Features a slot that is opened and closed by a vacuum-controlled flap
  • Description & Specifications
Related Products
Product Content Feedback