Learn More
Gilson™ MICROMAN™ Capillary Pistons
Made specifically for Gilson™ MICROMAN™ E pipets.Gilson™ MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.
Manufacturer:  Gilson™ F148714
 View more versions of this product
Catalog No. F148714G
Add to cart
Description
Description
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.
Specifications
Specifications
Gilson™ Microman™
50 to 250μL
CP250ST
Gilson™ Microman™
Box of 576
Sterile
PP (Polypropylene)