Learn More
Ajax Scientific Micrometer Caliper Products for Science Education Non-distribution product as customer accommodation.
Traditional English caliper with a 0-1 in. range
$42.25
Specifications
Includes Hardened Steel screw, zero adjustment and locking device
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S40505 Non-distribution product as customer accommodation. Products for Science Education
View Documents
Ajax Scientific
12078
Metric Unit Caliper
Each for $42.25
Add to cart
Description
Description
Calipers show decimal equivalents and have a satin finish with friction stop. Graduations on the English-Unit caliper are 0.001 in.. Metric Caliper has a 0-25mm range with 0.01mm graduations. Spindle locks for more precise comparison.
Specifications
Specifications
Hardened Steel screw, zero adjustment and locking device