Learn More
Micrometer Calipers
Manufacturer:  Walter Stern Inc.Supplier Diversity Partner 601B
 View more versions of this product
Catalog No. 12-126
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.001 in.
Positive spindle lock and friction stop, Forged Steel fork with hammer tone finish, Steel frame with Satin Chrome finished barrel with clearly graduated markings, padded Plastic Case
Micrometer Caliper
0 to 1 in.
English