Learn More
MilliporeSigma™ Supelco™ MicroMist™ Nebulizer, 0.6 mL/min. with EzyFit & EzyLok, Gas Flow: 0.7 L/min.
Suggested Gas Flow 0.7 L/min. Suggested Sample Uptake 0.6 mL/min.
$912.00
Specifications
For Use With (Application) Spectrometry
View More Specs
Products 2
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
11-100-7068
View Documents Promotion Details
MilliporeSigma™ Supelco™
GXAR4007FM06E1EA
GBC Scientific Integra XL, XM, XM2, XP, XMP
Each for $912.00
Add to cart
11-100-7067
View Documents Promotion Details
MilliporeSigma™ Supelco™
GXAR3507FM06ES1EA
Shimadzu 1000, 5000, 7000, 8000, 9000 Series
Each for $912.00
Add to cart
Description
Description
Specifications
Specifications
Spectrometry
SDS
Product Certifications
MicroMist is a trademark of Glass Expansion Pty Ltd