Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ MicroMop™ Adapters

These lightweight and durable, plastic string mop adapters work with a range of string mops and are available with the choice between a stainless steel handle attachment or an included plastic handle.

$85.00 - $130.00

Specifications

Color Orange
Disposable No
For Use With (Application) For Cleaning Large Surface Areas Quickly and Efficiently
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Type Material Autoclavable Price Quantity & Availability  
Catalog Number Mfr. No. Type Material Autoclavable Price Quantity & Availability  
19-041-813
View Documents
Micronova™
MH1
Attached Plastic Handle Fiberglass, Polypropylene Not Autoclavable
Each for $85.00
Add to cart
 
19-035-363A
View Documents
Micronova™
QDMH1
QD Attachment for Stainless Handle Stainless Steel, 304 Electropolished, Polypropylene Autoclavable
Each for $130.00
Add to cart
 
Description

Description

The Micronova MicroMop™ String Mop Adapters work with a variety of string mops, large and small, and are available with a choice of two handle options. The QDMH-1 adapter, with a Quick Disconnect handle attachment, connects to the stainless steel SSU series handles. The MH-1 adapter includes an attached, 60-inch plastic handle.

  • Orange polypropylene string mop holders
  • Lightweight and durable
  • Holds a range of string mops
  • QDMH-1 Adapter features a Stainless Steel, Quick Disconnect Handle Attachment
  • MH-1 Adapter includes an attached 60-inch plastic coated handle
  • Autoclavable
Specifications

Specifications

Orange
No
For Cleaning Large Surface Areas Quickly and Efficiently
Non-sterile
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit