Micronova™ Cleanroom Mops Special Tools and Accessories Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Micronova™ ITC6911IR
 View more versions of this product
Catalog No. 19-130-4802
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Isolators, containment and bio hoods in cleanroom and buffer areas
    Width (English) 9 in.
    Autoclavable Yes
    Material Fabric
    Product Type Mop Isolator Hood
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback