Micronova™ Cleanroom Pens Available on GSA/VA Contract for Federal Government customers only.
Low-sodium ink
Manufacturer: Micronova™ PEN01TIR
 View more versions of this product
Catalog No. 19-166-580
  Check Availability
Add to Cart
    Specifications
    Product Type Cleanroom Pen
    Color Black
    For Use With (Application) Marks on cold or wet surfaces, plastic bags, or disposable labware and marks will not smear on reusable glass, metal, or porcelain labware
    Material (Barrel) Polypropylene
    Material (Internal) Low Sodium ink
    Quantity 100 Case
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback