Learn More
Micronova™ CRV Vinyl Cleanroom Tape - 3/8 in. wide
SDP
Supplier:  Micronova™ CRV38YE
 View more versions of this product
Catalog No. 19-161-233
Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
PPP-T-66E, CID A-A 1689A
Yellow
Tape
Cleanroom Vinyl tape
For sealing curved and irregular surfaces, aisle marking, labeling, clean packaging, color coding
36 yd.
0.37 in.
Rubber, Synthetic
Matte
Vinyl
Non-sterile
36 yd. x 0.37 in. (32.91 m x 0.95 cm)
32.91 m
0.95 cm
Product Suggestions
Product Suggestions