Learn More
Micronova™ Double-Coated Cleanroom Tape
SDP
Supplier:  Micronova™ 1TRDC
 View more versions of this product
Catalog No. 19-058-390
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Made of PVC film coated with a long-lasting acrylic adhesive on each side, this linerless double-coated film tape is designed for cleanroom applications.
•    Differential adhesive eliminates the need to remove and discard liner material, so there is less waste
•    Adhesive bonds with varied materials, including metals, plastics, and glass
•    Extremely resistant to solvents and UV
Order Info
Wound on a plastic core in 36 yd. (33m) rolls
Compliance
Meets or exceeds UU-T-91E, Type 1
Product Suggestions
Product Suggestions