Learn More
Micronova™ Double-Coated Cleanroom Tape
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-058-391
View Documents
Micronova™
12TRDC
N/A 0.5 in. x 36 yd. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-058-390
View Documents
Micronova™
1TRDC
N/A 1 in. x 36 yd. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Made of PVC film coated with a long-lasting acrylic adhesive on each side, this linerless double-coated film tape is designed for cleanroom applications.
  • Differential adhesive eliminates the need to remove and discard liner material, so there is less waste
  • Adhesive bonds with varied materials, including metals, plastics, and glass
  • Extremely resistant to solvents and UV
Order Info
Wound on a plastic core in 36 yd. (33m) rolls
Compliance
Meets or exceeds UU-T-91E, Type 1