Micronova™ EconoMop™ Foam Mop Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Micronova™ ECM122318
 View more versions of this product
Catalog No. 19-130-4841
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Ideal for applying cleaning agents and disinfectants in controlled environments
    Length (English) 3 in.
    Width (English) 18 in.
    Includes Yellow foam brick type sponge head
    Material Foam
    Product Type Foam Mop
  • Description & Specifications
Related Products
Product Content Feedback