Learn More
Micronova™ Industrial-Grade Vinyl Cleanroom Tape
For light industrial use, clean construction and cleanroom support
Manufacturer:  Micronova™ EGVFP12WH/DEL
 View more versions of this product
Catalog No. 19-120-3265
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
White
0.5 in. x 36 yd.
36 yd.
0.5 in.
Vinyl
Each
Complies with Federal Specifications PPP-T-66E and CID A-A 1689A
Aisle marking, labeling, batching, clean-build projects
33m
Tape
Writeable surface cleanroom tape
No