Micronova™ M-Zone™ MegaSorb™ Mops

Designed for cleaning and picking up liquids in controlled and critical environments

Manufacturer: Micronova™ MZCRMS1112IR

 View all options for this product

Catalog No. 19-065-109


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color White
    Autoclavable Yes
    Material Polyester, Nylon
    Product Type M-zone MegaSorb™ Mop; Gamma irradiated
    Disposable No
    For Use With (Application) Strong disinfectants, floors
    Size 10 oz.

    • Made from microfiber tube which is both low-linting and highly absorbent
    • Tufted strands comprised of polyester and nylon filaments work to pick up tiny particles
    • For use with strong disinfectants
    • Able to be laundered, autoclaved, gamma-irradiated, and re-used
    • Can be used with Micronova MH-1 Mop Handle or attached to the Micronova universal stainless steel handle via the Micronova QDMH-1 Mop Adapter

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit