Learn More
Micronova™ MicroMop™ Fabric Mops Available on GSA/VA Contract for Federal Government customers only.
Highly efficient floor mops for cleaning large surface areas quickly
Manufacturer:  Micronova™ CRM6
 View more versions of this product
Catalog No. 19-035-364C
  Check Availability
Add to cart
Requires : Quick-disconnect MicroMop adapter (Cat. No. 19-035-363B) and extendable or single-length stainless-steel handles (sold separately).
Specifications Description & Specifications
Specifications
Yes
MicroMop™
Used for ISO class 3 to 8 cleanrooms, cleaning large surface areas quickly, floor
Polyester
No
Accessories