Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ MegaTape™ Polyethylene Cleanroom Tape
BioPharma

A medium density, polyethylene tape with hi-tack/clean-peel acrylic adhesive, good conformability and excellent resistance to moisture, oils and weathering. Ideal for clean construction and maintenance projects.

$23.55 - $662.00

Specifications

Adhesive Type Acrylic
Autoclavable Not Autoclavable
Color White
For Use With (Application) Ideal for Clean Construction and Maintenance Projects
Length (Metric) 33 m
View More Specs

Products 3
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
19-045-220 BioPharma
View Documents
Micronova™
MTP12WH
12.70 mm 0.5 in.
Each for $23.55
Case of 72 EA for $1,269.00
Add to cart
 
19-045-230 BioPharma
View Documents
Micronova™
MTP2WH
50.8 mm 2 in.
Case of 24 for $662.00
Add to cart
 
19-095-020 BioPharma
View Documents
Micronova™
MTP1WH
25.4 mm 1 in.
Case of 36 for $634.00
Add to cart
 
Description

Description

The Micronova MegaTape™ Polyethylene Cleanroom Tape, with hi-tack acrylic adhesive, provides superior sealing abilities and removes cleanly with no adhesive residue. This tape is low outgassing, has good conformability, similar to vinyl tapes, and shows excellent resistance to UV, moisture, acids, alkalis, oils, abrasion and weathering.

Specifications

Specifications

Acrylic
White
33 m
Polyethylene
Non-sterile
60°F to 80°F
Not Autoclavable
Ideal for Clean Construction and Maintenance Projects
36 yd.
Cleanroom Tape
16°C to 27°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit