Learn More
Micronova™ NovaPoly™ Cleanroom Mop
Polyester-covered foam for critical applications and sterile support areas
Manufacturer:  Micronova™ PH6154IR
Catalog No. 19-045-210
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Minimizes particulation
  • Can be laundered, autoclaved, reused
  • Irradiated
  • 14 in. wide
Specifications
Specifications
Safety
Yes
Cleanroom Mop
14 in.
Sterile
Polyester
For critical applications and sterile support areas