Micronova™ NovaPoly Sponge
Polyester covered urethane foam
$12.02 - $435.10
Specifications
Autoclavable Yes
Disposable No
For Use With Application Ideal for lapping and polishing in wafer manufacture and general wiping
For Use With Equipment Workstations, laminar flow benches, walls, windows
Material Polyester
View More Specs
  • Products 5
  • Description & Specifications
Products
Catalog Number Mfr. No. Dimensions (L x W) Product Type Sterile Thickness Price Quantity & Availability    
19-035-375A
Available on GSA/VA Contract for Federal Government customers only. 
Micronova Mfg.
SP6-33
3 x 3 in. (7.6 x 7.6cm) NovaPoly™ sponge No 0.75 in. Each for $13.14
Check Availability
Add to Cart
19-035-375B
Available on GSA/VA Contract for Federal Government customers only. 
Micronova Mfg.
SP6-46
4 x 6 in. (10.2 x 15.24cm) NovaPoly™ sponge No 0.5 in. Each for $12.02
Check Availability
Add to Cart
19-035-375C
Available on GSA/VA Contract for Federal Government customers only. 
Micronova Mfg.
SP6-745
7 x 4.5 in. (17.8 x 11.4cm) NovaPoly™ sponge No 0.5 in. Each for $12.98
Check Availability
Add to Cart
19-035-375D
Available on GSA/VA Contract for Federal Government customers only. 
Micronova Mfg.
SP6-363
3 x 6 in. (7.6 x 15.24cm) NovaPoly™ sponge No 3 in. Each for $23.08
Check Availability
Add to Cart
19-045-211
Available on GSA/VA Contract for Federal Government customers only. 
Micronova Mfg.
SP6-46IR
4 x 6 in. (10.2 x 15.24cm) NovaPoly™ sponge Irradiated Yes 0.5 in. Each for $435.10
Check Availability
Add to Cart
Descriptions & Specifications
Related Products