Micronova™ PadMop™ Sponge Mop Available on GSA/VA Contract for Federal Government customers only.
For use in pharmaceutical production or aseptic processing areas
Manufacturer: Micronova™ SLP618IR
 View more versions of this product
Catalog No. 19-165-648
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Pharmaceutical production or aseptic processing areas, floors, walls, ceilings
    Length (English) 18 in.
    Autoclavable Yes
    Material Polyester, Urethane Foam
    Product Type PadMop™ Sponge Mop
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback