Micronova™ PocketMop Flat Head Irradiated Mops Available on GSA/VA Contract for Federal Government customers only.
Used for controlled environments
Manufacturer: Micronova™ PKMAW18IR
 View more versions of this product
Catalog No. 19-045-255
  Check Availability
Add to Cart
    Specifications
    Disposable No
    For Use With (Application) Floors, walls, ceilings
    Length (English) 18 in.
    Size 18 in.
    Autoclavable Yes
    Material White Microfiber
    Product Type Irradiated Flat Head Mop
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback