Micronova™ Polyester SnapMop™ Mop Covers Available on GSA/VA Contract for Federal Government customers only.

Dries fast and weighs little. Micronova™ Polyester SnapMop™ Mop Covers are are washable, autoclavable, and reusable.

Manufacturer: Micronova™ MZSM5618

 View more versions of this product

Catalog No. 19-305-732

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Application) Cleaning floors, walls, ceilings in clean and controlled environments
    Length (English) 18 in.
    Width (English) 2 in.
    Dimensions (L x W) 46 x 5cm
    Quantity 24Pack
    Autoclavable Yes
    Material NovaPoly™
    Model MZSM56-18
    Disposable No
    Product Type SnapMop Mop Head
    Sterile No

    • Holds up to strong disinfectants and steam sterilization
    • Launderable
    • 100% Polyester

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit