Learn More
Micronova™ PolyMitt™ Wiper
Combines versatility of wipe with security of glove
Manufacturer:  Micronova™ MW6IR
 View more versions of this product
Catalog No. 19-166-661
Add to cart
Description
Description
  • 100% high-quality polyester mitt fits comfortably over gloved hand for cleaning ease around carts, shelving, tanks, tubing and other hard-to-reach areas
  • Allows more dexterity in cleaning procedures as the mitt is virtually attached to operator's hand
  • Can also be used as a protective layer over standard gloves in some clean, build and maintenance applications
  • Available in two materials and with foam interior for increased sorbency
  • Universal size
Specifications
Specifications
Cleaning ease around carts, shelving, tanks, tubing and other hard to reach areas, used as a protective layer over standard gloves
Polyester
PolyMitt Mitt Wipes without sponge