Micronova™ PowerHead™ Disposable Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Available in several different types and densities of foam and material coverings
Manufacturer: Micronova™ PHU110
 View more versions of this product
Catalog No. 19-041-807A
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Designed for use in cleanroom support areas
    Width (English) 10 in.
    Autoclavable Yes
    Material Dense Urethane Foam
    Product Type PowerHeads™ Urethane Dense Foam Mop Head
    Disposable Yes
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback