Micronova™ PowerHead™ Disposable Mop Heads
Available in several different types and densities of foam and material coverings
Manufacturer: Micronova™ PHU144
 View all options for this product
Catalog No. 19-035-372B
  Check Availability
Add to Cart
    Autoclavable Yes
    Material Ether Foam
    Product Type PowerHeads™ Ether Foam Mop Head
    Sterile Yes
    Disposable Yes
    For Use With (Application) Dense and absorbent for use in aseptic and sterile support areas
    Width (English) 14 in.
    Suitable for use on floors and walls. Complete mop consists of a PowerHead disposable mop head, a PowerHead handle (sold separately) and an extendible or single-length stainless-steel handle (sold separately).
    PowerHead mop types include:
    Yellow foam
    • Heavy-duty
    • Two distinct cleaning surfaces
    Dense Urethane Foam
    • Disposable sponge mop head for cleanroom support areas
    • Two distinct cleaning surfaces
    • Withstands strong disinfectants
    Ether and Dense Ether Foam
    • Ideal for applying detergents and disinfectants
    • Dense absorbent foam for aseptic and sterile support areas
    • Two distinct cleaning surfaces
    Wide-Pore Foam
    • Economical, absorbent foam for pharmaceutical and sterile support areas
    • Suited for detergent and disinfectant applications
    • Two distinct cleaning surfaces
    NovaPoly™ Covered Foam
    • Polyester covered to minimize particulations
    • For critical applications, pharmaceutical and sterile support facilities
  • Description & Specifications
Related Products
Product Content Feedback