Micronova™ SlimLine™ Cleanroom Mop: Covers
For floor and wall cleaning
Manufacturer: Micronova™ SLSC6814
 View all options for this product
Catalog No. 19-041-46
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Required Accessories : Mop adapters and extendible or standard stainless-steel handle (sold separately).
    Autoclavable Yes
    Material MicroFiber
    Product Type SlimeLine™ NovaLite Cover
    For Use With (Application) Suitable for use with strong disinfectants and repeated autoclaving
    For Use With (Equipment) SlimLine cleanroom mop
    Width (English) 14 in.
    NovaPoly™ Slipcovers
    • Launderable and reusable, and easy to change
    • Saturate thoroughly for sterilizing purposes
    • Suitable for use with strong disinfectants and repeated autoclaving
    Polymesh Cover
    • Urethane foam interior
    • For heavy duty cleaning tasks, high traffic areas, staining and chemical residues
    • Monofilament mesh offers increased abrasion and minimal particulation
  • Description & Specifications
Related Products
Product Content Feedback