Learn More
Micronova™ SST™ Striped Safety Tape
A striped, laminated vinyl safety tape that is designed for delineating and color coding specific equipment and areas within cleanrooms and controlled environments. Available in black/yellow, red/white, and black/white for differentiating marks.
Supplier: Micronova™ SST182BLWH
Description
For delineating and color coding specific equipment and areas
The Micronova SST™ Striped Safety Tape is a striped, pressure-sensitive tape with rubber adhesive that is ideal for marking fire equipment, exits, hazardous areas and first aid points within cleanrooms and controlled environments. The laminated vinyl is scuff resistant and holds up well to solvents, humidity and oil.
- Laminated vinyl film with rubber adhesive
- Cleanroom processed, pressure sensitive and scuff resistant
- Designed for delineating and color coding in cleanrooms and controlled environments
- Ideal for marking fire equipment, exits, hazardous areas and first aid points
- Available in black/yellow, red/white, and black/white stripes
- Holds up well to solvents, humidity and oil
- Wound on a 3-inch plastic core
Order Info
Available in 1, 2 or 3 in. widths.
Specifications
Not Autoclavable | |
Black, White | |
Cleanroom Tape | |
Delineating and color-coding specific equipment and areas | |
16°C to 27°C | |
18 yd. | |
2 in. |
Rubber | |
Vinyl | |
Non-sterile | |
60°F to 80°F | |
16.5 m x 50.8 mm (18 yd. x 2 in.) | |
16.5 m | |
50.8 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.