Learn More
Micronova™ T-Mop™ Cleanroom Mops with Covers Available on GSA/VA Contract for Federal Government customers only.
Low-profile, wide-surface mops with covers
Manufacturer:  Micronova™ T118
 View more versions of this product
Catalog No. 19-035-368G
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
T-Mop™ Cleanroom Mop
Ideal for applying disinfectants in sterile environments or for general cleaning in ISO class 3 to 8 cleanroom environments
NovaPoly™ slipcover
Polyester, Urethane Foam
Yes
18 in.