Learn More
Micronova™ Vinyl Tape
Products 3
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19-061753
View Documents
Micronova™
2WH47PC
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-061749
View Documents
Micronova™
1WH47PC
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-062684
View Documents
Micronova™
12WH47PC
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Vinyl tape features nontransferring adhesive and a 3 in. (7.6cm) poly core.
  • Resists most acids, alkalis, and bacteria
  • Tape easily conforms to curved and irregular surfaces
  • Matte finish can be written on with a cleanroom pen
Order Info
Available in a variety of colors and sizes for coding, identification, sealing, labeling, or aisle marking.
Roll length, 36 yards (32.9m)