Micronova™ Yellow Plastic Buckets and Wringers Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Micronova™ W1

 View more versions of this product

Catalog No. 19-035-360A

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 24 to 36 oz.
    Capacity (Metric) 0.7 to 1.06L
    Capacity 24 to 36 oz., 0.7 to 1.06L
    Color Yellow
    For Use With (Application) Cleanroom support applications and general janitorial use
    For Use With (Equipment) Buckets
    Material Plastic
    Product Type Wringer
    Quantity Each

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit