Learn More
Vileda Professional MicroOne Pocket Mop
MicroOne Mops offer microfiber cleaning performance in a disposable format, with a sturdy feel of a durable mop.
Manufacturer: Vileda Professional 159917
Description
- Microfiber cleaning performance in a disposable mop
- Fast, stubborn dirt removal and spill pick-up
- Double the absorbency of compatible mops
- Improved fit onto the frame
- Fits CombiSpeed Pro Frame in addition to other common industry frames
- 4 Colors available
- Low friction and superb ergonomics
- Compliant with a wide range of cleaning solutions
- Single use concept reduces the risk of cross-contamination
Specifications
Microfiber | |
Disposable Microfiber Pocket Mop | |
16 in. | |
Green | |
Bucket or pre-prepared method |
Non-sterile | |
25/Pk., 5 Pk./Cs. | |
Not Autoclavable | |
Yes | |
CombiSpeed Frame (143572) Telescopic handle (119966) |