Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Microphthalmia (Transcription Factor) (MiTF) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Microphthalmia (Transcription Factor) (MiTF) Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS772P0

 View more versions of this product

Catalog No. MS772P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Microphthalmia (Transcription Factor) (MiTF) Ab-2, Mouse Monoclonal Antibody.

Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52kDa and 56kDa, while the longer Mi form runs as a cluster of bands at 60-70kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.

Host Species: Mouse

Clone: D5

Isotype: IgG1/κ

Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.

Epitope: N-terminal

Immunogen: N-terminal fragment of human Mi protein

Molecular Weight: 52-56kDa doublet

Positive Control: 501 Mel cells, Melanoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Microphthalmia (Transcription Factor) (MiTF) Ab-2
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin)
D5
Unconjugated
Concentrated
N-terminal fragment of human Mi protein
100μL
Transcription Regulation
Primary
Canine, Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit