Learn More
Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Integrate these microplate carriers with the microplate swinging bucket rotor.
$1249.00
Specifications
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
75-003-625 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
75003625
Sealed Microplate Carriers Each for $1,249.00
Add to cart
Description
Description
Integrate Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor to add flexibility to the swinging-bucket rotor.
Includes:
Sealed carriers include trays and lids (set of 2). Replacement O-rings for lids are offered as a set of four, with grease.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Biocontainment certified by CAMR™, HPA, Porton Down, U.K.