Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
$2700.00
Products 1
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
50950379
View Documents
Thermo Scientific™
24072800
Multiskan Verification Plate with 8 wavelengths
  • NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate
  • Certificate of Calibration
  • USB memory device including SkanIt™ software sessions and reference files
  • Carrying case for Multiskan™ Verification Plate
Each for $2,700.00
Add to cart
Description
Description
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation. 
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
For Research Use Only. Not for use in diagnostic procedures.