Learn More
Research Products International CorpSupplier Diversity Partner 96 Well Plate Guide, Horizontal, 10 per Package DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 247815
Bright color lanes in horizontal orientation clearly define rows on assay plates. Helps reduce fatigue and errors when pipetting multiple samples into plates. Reusable cards fit most standard well plates.
Catalog No. 50-476-532