Learn More
Valutek™ Microporous Coverall with Full Zipper, Elastic Cuff & Ankle
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- All coveralls have a flap over zipper closure, elastic wrist, and ankle cuffs design
- Available with either a single ply shirt collar or with an elastic hood and attached anti-skid PVC sole boot covers
- The coveralls are highly breathable and provide the operators with comfortable full body coverage protection
- All Valutek™ coveralls are tested and manufactured in ISO compliant facilities under Valutek™ inspection and stringent process control to ensure Valutek™ quality standard and product specification compliance
- This product is recommended for either stringent industrial or demanding cleanroom applications
- This coverall is ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment as well as other critical areas that require high contamination control and cleanliness
Specifications
Specifications
Size | Small |
Disposable | Single-use |
Cuff Style | Elastic |
Closure Type | Zipper |
Sterility | Non-sterile |
Resistance | Non-toxic Particulates |