Learn More
Thermo Scientific™ Microprotein Reagent
Test for protein in urine and cerebrospinal fluid with Thermo Scientific™ Microprotein Reagent.
$67.70
Specifications
CE Marker Yes
Stability Stable Liquid
State at Room Temperature Liquid
View More Specs
Products 1
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
TR50001
Accessories View Documents
Microprotein - Benzethonium Chloride (2-Part Rgts)
Pack of 2 for $67.70
Add to cart
Description
Description
Test for protein in urine and cerebrospinal fluid with Thermo Scientific™ Microprotein Reagent, a stable liquid that minimizes laboratory reagent waste.
Protein may be present in the urine under non-pathological conditions such as pregnancy, dehydration and exercise. Pathologic increases are seen with severe burns, nephrotic syndrome, multiple myeloma, heavy metal poisoning, diabetic nephropathy, pyelonephritis and preeclampsia. Protein will be elevated in cerebrospinal fluid with meningitis, encephalitis, brain tumors, intracranial hemorrhage, traumatic injury and poliomyelitis.
  • Intended for use with both manual and automated systems
Specifications
Specifications
Yes
Stable Liquid
Liquid