Learn More
Fisherbrand™ Microscope Cover Glasses: Squares
SCI_ED
Packed for easy dispensing; clean and ready-to-use.
 View more versions of this product
Catalog No. S17522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Uniform thickness and size
  • Noncorrosive, nonfogging, and free from bubbles, scratches, and striations
Order Info
Sold as One Each or Case of 10.
Specifications
Specifications
Cover Glass
18 mm
0.17 to 0.25 mm
Microscopes
Square
18 mm
18 x 18 mm