Learn More
Simport™ Scientific Microscreen™ I Small Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeve
Microscreen™ I Small Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeve are designed for small and delicate specimens.
Supplier:  Simport™ Scientific M52112SL
 View more versions of this product
Catalog No. 23-045-407
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Microscreen™ cassette is made of a strong nylon mesh.
  • Designed to handle small, fragile biopsies.
  • Nylon mesh has 1,500 square openings (300 μm each).
  • 8 x 11 mm slots in base and lid to help cassette sink rapidly.
  • Sturdy nylon mesh improves fluid exchange by reducing surface tension of reagents.
  • Accelerates fixation/preparation of tissue samples.
  • Eliminates the need for pads, sponges, nylon or lens paper bags.
  • Reduces the number of incidents involving non-fixed or insufficiently fixed tissues.
  • Inner chamber design for greater biopsy protection.
  • Large front tab for easier lid opening.
  • Hinge design configuration allows for multiple opening and closing.
  • In convenient Quickload™ sleeves, suitable for automated printers.
Specifications
Specifications
45°
Aqua
Acetal
Cassette