Learn More
American Educational Products Microslide™ Viewer Products for Science Education
Specially designed teaching tool for viewing microslides. The viewer does not require electricity or batteries and can be used outdoors as well as the classroom.
Manufacturer:  American Educational Products T001
Catalog No. S19220
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
  • Used to view a microslide, which has 8 related 35mm images as photographed through a microscope
  • White reflector provides sufficient light for a bright image in normal light of a classroom
  • Arrows and call-outs help students locate important features being studied
  • Microslide is mounted in a clear plastic holder that protects both its sides
  • Precision instrument, made in the USA
  • Unconditionally guaranteed against breakage for 5 years
Specifications
Specifications
Education
22.86cm
5x
5 Years
9 in.
5x
Used to view slides photographed through microscopes