Thermo Scientific™ MicroTest™ Tubes Available on GSA/VA Contract for Federal Government customers only.

Maintain viability of viruses chlamydiae, mycoplasma and urealasma with top quality transport and storage media that comes in four versatile formulations.

Manufacturer: Thermo Scientific™ R12503

 View more versions of this product

Catalog No. 23-499-806

  Check Availability
Add to Cart

    Specifications

    Closure Color Blue
    Shelf Life 18 months
    For Use With viruses, chlamydiae, Ureaplasmas, and Mycoplasmas
    Transport Temperature -25° to 8°C
    Fill Volume 3mL/Tube
    Description M4 Tube w/o Beads
    Packaging Packaged in partitioned boxes.
    Quantity 72 Tubes/Pk.
    Storage Requirements -25°C or 2-8°C
    Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology

    Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.

    Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

    MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

    MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae. MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

    MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

    MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.

    Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

    Trusted reliable results

    • Stringent quality control, including cellular toxicity on each lot number
    • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
    Compatibility and viability
    • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
    • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
    Convenient, accommodating format
    • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
    • Individual transport media tubes or complete kits offer flexibility and versatility
    Safe, sturdy packaging
    • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
    • Kits include leak-resistant polypropylene bag for safe sample transport

    Not all products are available for sale in all territories.

    Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit