Thermo Scientific™ MicroTest™ Universal Plus Kits Available on GSA/VA Contract for Federal Government customers only.

Maintain organism viability with kits including transport media and two traditional-tipped swabs plus an envelope with both traditional and micro-tipped polyester swabs.

Manufacturer: Thermo Scientific™ R12576

 View more versions of this product

Catalog No. 23-246-397

  Check Availability
Add to Cart

    Specifications

    Closure Color Red
    Shelf Life 18 months
    For Use With viruses, chlamydiae
    Transport Temperature 2° to 30°C
    Fill Volume 3mL/Tube
    Description M4RT Universal Plus Kit
    Quantity 100 Kits/Pk.
    Shaft Plastic and stainless steel
    Swab Configuration Traditional tipped and micro tipped
    Storage Requirements Room temperature
    Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology

    Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging. Thermo Scientific™ MicroTest™ Universal Plus Kits come in two media formulations and include two plastic shaft traditional-tipped polyester swabs plus an additional envelope with one plastic shaft traditional-tipped polyester swab and one stainless steel shaft micro-tipped polyester swab and a 6 x 9 in. zip-seal bag for safe transport.

    Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

    MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

    MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

    MicroTest Universal Plus Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads, an envelope with two plastic shaft traditional-tipped polyester swabs plus an envelope with one plastic shaft, traditional-tipped polyester swab and one stainless steel shaft micro-tipped polyester swab, and a 6 x 9 in. zip-seal bag.

    Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

    Trusted reliable results

    • Stringent quality control, including cellular toxicity on each lot number
    • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
    Compatibility and viability
    • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
    • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing.
    Convenient, accommodating format
    • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
    • Individual transport media tubes or complete kits offer flexibility and versatility
    Safe, sturdy packaging
    • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
    • Kits include leak-resistant polypropylene bag for safe sample transport

    Not all products are available for sale in all territories. Please inquire.

    Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit