Learn More
MTC Bio Microtubes, 1.5ml w/ green screw caps packed separately, 500/pk DFS Item
Manufacturer:  MTC Bio C2210
From the SureSeal family of sterile tubes you trust comes MTC Bio’s new 2.0mL screw cap microcentrifuge tubes. Made from ultra-clear polypropylene, with crisp printed graduations, SureSeal 2mL screw-cap tubes offer the best visualization of sample on the market today! May be centrifuged at up to 22,000xg. (standard non-sterile 1.5mL screw cap tubes are also available)
Catalog No. 50-112-7175