Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Scientific™ MicroTube™ Cap
$46.65
Specifications
Material | Polyethylene |
---|---|
Color | Natural |
Sterility | Non-sterile |
For Use With (Equipment) | Microtube Rack System |
Catalog Number | Mfr. No. | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Price | Quantity & Availability | |||||
11-842-78
|
National Scientific™
TN3346 08C |
8-Capstrips |
Pack of 120 for $46.65
Case of 5 PK for $192.40
|
|
|||||
11-842-80
|
National Scientific™
TN3346 12C |
12-Capstrips |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Specifications
Polyethylene | |
Natural | |
Non-sterile | |
Microtube Rack System |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title